Assay Details
Gene Symbol: LOC5573048 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654317.2 | XP_001654367.2 | 5 - 6 | 1453 | 76 | |
XM_021849041.1 | XP_021704733.1 | 6 - 7 | 1463 | 76 | ||
XM_021849042.1 | XP_021704734.1 | 6 - 7 | 1383 | 76 | ||
XM_021849043.1 | XP_021704735.1 | 6 - 7 | 1367 | 76 | ||
XM_021849044.1 | XP_021704736.1 | 7 - 8 | 1375 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
