Assay Details
Gene Symbol: LOC5573048 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654317.2 | XP_001654367.2 | 3 - 4 | 918 | 60 | |
XM_021849041.1 | XP_021704733.1 | 4 - 5 | 928 | 60 | ||
XM_021849042.1 | XP_021704734.1 | 4 - 5 | 848 | 60 | ||
XM_021849043.1 | XP_021704735.1 | 4 - 5 | 832 | 60 | ||
XM_021849044.1 | XP_021704736.1 | 5 - 6 | 840 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
