Assay Details
Gene Symbol: LOC5573048 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654317.2 | XP_001654367.2 | 1 - 2 | 616 | 76 | |
XM_021849041.1 | XP_021704733.1 | 2 - 3 | 626 | 76 | ||
XM_021849042.1 | XP_021704734.1 | 2 - 3 | 546 | 76 | ||
XM_021849043.1 | XP_021704735.1 | 2 - 3 | 530 | 76 | ||
XM_021849044.1 | XP_021704736.1 | 3 - 4 | 538 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
