Assay Details
Gene Symbol: LOC5573486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842254.1 | XP_021697946.1 | 1 - 2 | 1060 | 81 | |
XM_021842255.1 | XP_021697947.1 | 1 - 2 | 1060 | 81 | ||
XM_021842256.1 | XP_021697948.1 | 1 - 2 | 1059 | 81 | ||
XM_021842257.1 | XP_021697949.1 | 1 - 2 | 1059 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021842254.1, XM_021842255.1, XM_021842256.1, XM_021842257.1 transcripts.
Set Membership: |
![]() ![]() |
