Assay Details
Gene Symbol: LOC5573486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842254.1 | XP_021697946.1 | 2 - 3 | 4464 | 77 | |
XM_021842255.1 | XP_021697947.1 | 3 - 4 | 4374 | 77 | ||
XM_021842256.1 | XP_021697948.1 | 4 - 5 | 3485 | 77 | ||
XM_021842257.1 | XP_021697949.1 | 4 - 5 | 3467 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
