Assay Details
Gene Symbol: LOC23687783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011495178.2 | XP_011493480.1 | 2 - 3 | 497 | 71 | |
XM_021845517.1 | XP_021701209.1 | 2 - 3 | 290 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
