Assay Details
Gene Symbol: LOC23687777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854883.1 | XP_021710575.1 | 3 - 4 | 6574 | 79 | |
XM_021854884.1 | XP_021710576.1 | 4 - 5 | 6377 | 79 | ||
XM_021854885.1 | XP_021710577.1 | 4 - 5 | 6382 | 79 | ||
XM_021854886.1 | XP_021710578.1 | 3 - 4 | 5763 | 79 | ||
XM_021854887.1 | XP_021710579.1 | 3 - 4 | 5828 | 79 |
