Assay Details
Gene Symbol: LOC23687777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854883.1 | XP_021710575.1 | 7 - 8 | 8226 | 84 | |
XM_021854884.1 | XP_021710576.1 | 8 - 9 | 8029 | 84 | ||
XM_021854885.1 | XP_021710577.1 | 8 - 9 | 8034 | 84 | ||
XM_021854886.1 | XP_021710578.1 | 7 - 8 | 7415 | 84 | ||
XM_021854887.1 | XP_021710579.1 | 7 - 8 | 7480 | 84 |
