Assay Details
Gene Symbol: LOC23687777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854883.1 | XP_021710575.1 | 8 - 9 | 8416 | 59 | |
XM_021854884.1 | XP_021710576.1 | 9 - 10 | 8219 | 59 | ||
XM_021854885.1 | XP_021710577.1 | 9 - 10 | 8224 | 59 | ||
XM_021854886.1 | XP_021710578.1 | 8 - 9 | 7605 | 59 | ||
XM_021854887.1 | XP_021710579.1 | 8 - 9 | 7670 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
