Assay Details
Gene Symbol: LOC110678647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851784.1 | XP_021707476.1 | 2 - 3 | 557 | 83 | |
XM_021851785.1 | XP_021707477.1 | 2 - 3 | 790 | 83 | ||
XM_021851786.1 | XP_021707478.1 | 2 - 3 | 557 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021851784.1, XM_021851785.1, XM_021851786.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
