Assay Details
Gene Symbol: LOC110678647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851784.1 | XP_021707476.1 | 6 - 7 | 3759 | 69 | |
XM_021851785.1 | XP_021707477.1 | 6 - 7 | 3992 | 69 | ||
XM_021851786.1 | XP_021707478.1 | 6 - 7 | 3759 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
