Assay Details
Gene Symbol: LOC110678647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851784.1 | XP_021707476.1 | 9 - 10 | 4546 | 85 | |
XM_021851785.1 | XP_021707477.1 | 9 - 10 | 4779 | 85 | ||
XM_021851786.1 | XP_021707478.1 | 8 - 9 | 4462 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
