Assay Details
Gene Symbol: LOC5564786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649077.2 | XP_001649127.1 | 7 - 8 | 1330 | 82 | |
XM_021851923.1 | XP_021707615.1 | 7 - 8 | 1555 | 82 | ||
XM_021851924.1 | XP_021707616.1 | 7 - 8 | 1369 | 82 |
