Assay Details
Gene Symbol: LOC5568617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652399.2 | XP_001652449.1 | 5 - 6 | 861 | 95 | |
XM_011494738.2 | XP_011493040.1 | 5 - 6 | 783 | 95 | ||
XM_021847059.1 | XP_021702751.1 | 5 - 6 | 759 | 95 | ||
XM_021847060.1 | XP_021702752.1 | 5 - 6 | 690 | 95 | ||
XM_021847061.1 | XP_021702753.1 | 4 - 5 | 505 | 95 | ||
XM_021847062.1 | XP_021702754.1 | 4 - 5 | 568 | 95 |
