Assay Details
Gene Symbol: LOC5580331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844809.1 | XP_021700501.1 | 6 - 7 | 2903 | 93 | |
XM_021844810.1 | XP_021700502.1 | 4 - 5 | 1043 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
