Assay Details
Gene Symbol: LOC5580331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844809.1 | XP_021700501.1 | 5 - 6 | 2704 | 67 | |
XM_021844810.1 | XP_021700502.1 | 3 - 4 | 844 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
