Assay Details
Gene Symbol: LOC5571056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659448.2 | XP_001659498.2 | 8 - 9 | 1851 | 79 | |
XM_021852816.1 | XP_021708508.1 | 8 - 9 | 1883 | 79 | ||
XM_021852817.1 | XP_021708509.1 | 8 - 9 | 1624 | 79 | ||
XM_021852818.1 | XP_021708510.1 | 8 - 9 | 1636 | 79 | ||
XM_021852819.1 | XP_021708511.1 | 8 - 9 | 1521 | 79 | ||
XM_021852820.1 | XP_021708512.1 | 8 - 9 | 1558 | 79 | ||
XM_021852821.1 | XP_021708513.1 | 8 - 9 | 1595 | 79 | ||
XM_021852822.1 | XP_021708514.1 | 8 - 9 | 1504 | 79 | ||
XM_021852823.1 | XP_021708515.1 | 8 - 9 | 1545 | 79 | ||
XM_021852824.1 | XP_021708516.1 | 8 - 9 | 1851 | 79 | ||
XM_021852825.1 | XP_021708517.1 | 8 - 9 | 1853 | 79 |
