Assay Details
Gene Symbol: LOC5566966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651333.2 | XP_001651383.2 | 3 - 4 | 923 | 74 | |
XM_001651334.2 | XP_001651384.1 | 3 - 4 | 923 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
