Assay Details
Gene Symbol: LOC5566966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651333.2 | XP_001651383.2 | 4 - 5 | 1323 | 93 | |
XM_021845409.1 | XP_021701101.1 | 4 - 5 | 1326 | 93 | ||
XM_021845410.1 | XP_021701102.1 | 4 - 5 | 1326 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
