Assay Details
Gene Symbol: LOC5566966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651332.2 | XP_001651382.2 | 5 - 6 | 1464 | 60 | |
XM_001651333.2 | XP_001651383.2 | 6 - 7 | 1616 | 60 | ||
XM_021845409.1 | XP_021701101.1 | 6 - 7 | 1619 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
