Assay Details
Gene Symbol: LOC5574012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852676.1 | XP_021708368.1 | 2 - 3 | 3245 | 75 | |
XM_021852720.1 | XP_021708412.1 | 3 - 4 | 2041 | 75 | ||
XM_021852839.1 | XP_021708531.1 | 3 - 4 | 2186 | 75 |
