Assay Details
Gene Symbol: LOC5574012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852676.1 | XP_021708368.1 | 4 - 5 | 3599 | 91 | |
XM_021852720.1 | XP_021708412.1 | 5 - 6 | 2395 | 91 | ||
XM_021852783.1 | XP_021708475.1 | 4 - 5 | 1918 | 91 | ||
XM_021852839.1 | XP_021708531.1 | 5 - 6 | 2540 | 91 |
