Assay Details
Gene Symbol: LOC5574012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852676.1 | XP_021708368.1 | 5 - 6 | 3816 | 56 | |
XM_021852720.1 | XP_021708412.1 | 6 - 7 | 2612 | 56 | ||
XM_021852783.1 | XP_021708475.1 | 5 - 6 | 2135 | 56 | ||
XM_021852839.1 | XP_021708531.1 | 6 - 7 | 2757 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
