Assay Details
Gene Symbol: LOC5574012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852676.1 | XP_021708368.1 | 6 - 7 | 3975 | 65 | |
XM_021852720.1 | XP_021708412.1 | 7 - 8 | 2771 | 65 | ||
XM_021852783.1 | XP_021708475.1 | 6 - 7 | 2294 | 65 | ||
XM_021852839.1 | XP_021708531.1 | 7 - 8 | 2916 | 65 |

More Information
Set Membership: |
![]() ![]() |
