Assay Details
Gene Symbol: LOC5574012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852676.1 | XP_021708368.1 | 7 - 8 | 4148 | 94 | |
XM_021852720.1 | XP_021708412.1 | 8 - 9 | 2944 | 94 | ||
XM_021852783.1 | XP_021708475.1 | 7 - 8 | 2467 | 94 | ||
XM_021852839.1 | XP_021708531.1 | 8 - 9 | 3089 | 94 |
