Assay Details
Gene Symbol: LOC5574012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852676.1 | XP_021708368.1 | 9 - 10 | 4386 | 71 | |
XM_021852720.1 | XP_021708412.1 | 10 - 11 | 3182 | 71 | ||
XM_021852783.1 | XP_021708475.1 | 9 - 10 | 2705 | 71 | ||
XM_021852839.1 | XP_021708531.1 | 10 - 11 | 3327 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
