Assay Details
Gene Symbol: LOC5572794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654145.2 | XP_001654195.2 | 1 - 2 | 400 | 89 | |
XM_021846989.1 | XP_021702681.1 | 1 - 2 | 397 | 89 | ||
XM_021846990.1 | XP_021702682.1 | 1 - 2 | 404 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021846990.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
