Assay Details
Gene Symbol: LOC5572794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654145.2 | XP_001654195.2 | 3 - 4 | 724 | 102 | |
XM_021846989.1 | XP_021702681.1 | 3 - 4 | 721 | 102 | ||
XM_021846990.1 | XP_021702682.1 | 3 - 4 | 732 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
