Assay Details
Gene Symbol: LOC5568747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658077.2 | XP_001658127.2 | 4 - 5 | 1013 | 104 | |
XM_021842362.1 | XP_021698054.1 | 4 - 5 | 1031 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
