Assay Details
Gene Symbol: LOC5568747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658077.2 | XP_001658127.2 | 6 - 7 | 1392 | 114 | |
XM_021842362.1 | XP_021698054.1 | 6 - 7 | 1410 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
