Assay Details
Gene Symbol: LOC5567951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657717.2 | XP_001657767.2 | 1 - 2 | 380 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5567951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657717.2 | XP_001657767.2 | 1 - 2 | 380 | 61 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |