Assay Details
Gene Symbol: LOC5575771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849104.1 | XP_021704796.1 | 2 - 3 | 346 | 91 | |
XM_021849105.1 | XP_021704797.1 | 2 - 3 | 268 | 91 | ||
XM_021849107.1 | XP_021704799.1 | 2 - 3 | 346 | 91 | ||
XM_021849108.1 | XP_021704800.1 | 2 - 3 | 346 | 91 | ||
XM_021849109.1 | XP_021704801.1 | 2 - 3 | 346 | 91 | ||
XM_021849110.1 | XP_021704802.1 | 2 - 3 | 346 | 91 | ||
XM_021849111.1 | XP_021704803.1 | 2 - 3 | 346 | 91 | ||
XM_021849112.1 | XP_021704804.1 | 2 - 3 | 346 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
