Assay Details
Gene Symbol: LOC5575771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849109.1 | XP_021704801.1 | 9 - 10 | 2449 | 99 | |
XM_021849111.1 | XP_021704803.1 | 8 - 9 | 2437 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
