Assay Details
Gene Symbol: LOC23687749 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021856114.1 | XP_021711806.1 | 4 - 5 | 3303 | 70 | |
XM_021856123.1 | XP_021711815.1 | 4 - 5 | 3287 | 70 | ||
XM_021856129.1 | XP_021711821.1 | 4 - 5 | 3304 | 70 | ||
XM_021856136.1 | XP_021711828.1 | 4 - 5 | 3229 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
