Assay Details
Gene Symbol: LOC5564296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648595.2 | XP_001648645.2 | 2 - 3 | 263 | 89 | |
XM_021851417.1 | XP_021707109.1 | 2 - 3 | 250 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
