Assay Details
Gene Symbol: LOC5574541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655144.2 | XP_001655194.1 | 8 - 9 | 1760 | 65 | |
XM_021843576.1 | XP_021699268.1 | 7 - 8 | 1823 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
