Assay Details
Gene Symbol: LOC5574541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655144.2 | XP_001655194.1 | 3 - 4 | 739 | 69 | |
XM_021843576.1 | XP_021699268.1 | 3 - 4 | 745 | 69 |

More Information
Set Membership: |
![]() ![]() |
