Assay Details
Gene Symbol: LOC5575906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844424.1 | XP_021700116.1 | 7 - 8 | 1599 | 82 | |
XM_021844425.1 | XP_021700117.1 | 7 - 8 | 1596 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
