Assay Details
Gene Symbol: LOC5569754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848396.1 | XP_021704088.1 | 2 - 3 | 185 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021848396.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
