Assay Details
Gene Symbol: LOC5569754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848396.1 | XP_021704088.1 | 4 - 5 | 4092 | 80 | |
XM_021848397.1 | XP_021704089.1 | 4 - 5 | 4010 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
