Assay Details
Gene Symbol: LOC5569754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848396.1 | XP_021704088.1 | 6 - 7 | 4550 | 85 | |
XM_021848397.1 | XP_021704089.1 | 6 - 7 | 4468 | 85 | ||
XM_021848398.1 | XP_021704090.1 | 3 - 4 | 556 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
