Assay Details
Gene Symbol: LOC5569754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848396.1 | XP_021704088.1 | 8 - 9 | 4997 | 113 | |
XM_021848397.1 | XP_021704089.1 | 8 - 9 | 4915 | 113 | ||
XM_021848398.1 | XP_021704090.1 | 5 - 6 | 1003 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
