Assay Details
Gene Symbol: LOC5572482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660380.2 | XP_001660430.2 | 2 - 3 | 528 | 92 | |
XM_021850427.1 | XP_021706119.1 | 3 - 4 | 536 | 92 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5572482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660380.2 | XP_001660430.2 | 2 - 3 | 528 | 92 | |
XM_021850427.1 | XP_021706119.1 | 3 - 4 | 536 | 92 |