Assay Details
Gene Symbol: LOC5572482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660380.2 | XP_001660430.2 | 11 - 12 | 2142 | 136 | |
XM_021850427.1 | XP_021706119.1 | 12 - 13 | 2150 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() |
