Assay Details
Gene Symbol: LOC5572482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660380.2 | XP_001660430.2 | 10 - 11 | 2023 | 103 | |
XM_021850427.1 | XP_021706119.1 | 11 - 12 | 2031 | 103 |

More Information
Set Membership: |
![]() ![]() |
