Assay Details
Gene Symbol: LOC5572482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660380.2 | XP_001660430.2 | 9 - 10 | 1855 | 93 | |
XM_021850427.1 | XP_021706119.1 | 10 - 11 | 1863 | 93 |
