Assay Details
Gene Symbol: LOC5572482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660380.2 | XP_001660430.2 | 8 - 9 | 1737 | 83 | |
XM_021850427.1 | XP_021706119.1 | 9 - 10 | 1745 | 83 |
