Assay Details
Gene Symbol: LOC5578830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842184.1 | XP_021697876.1 | 4 - 5 | 1027 | 76 | |
XM_021842185.1 | XP_021697877.1 | 4 - 5 | 608 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
