Assay Details
Gene Symbol: LOC5564252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839017.1 | XP_021694709.1 | 6 - 7 | 1572 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5564252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839017.1 | XP_021694709.1 | 6 - 7 | 1572 | 78 |
Set Membership: |
![]() ![]() ![]() ![]() |