Assay Details
Gene Symbol: LOC5564121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855336.1 | XP_021711028.1 | 4 - 5 | 1517 | 61 | |
XM_021855337.1 | XP_021711029.1 | 3 - 4 | 1497 | 61 | ||
XM_021855338.1 | XP_021711030.1 | 4 - 5 | 1512 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
